Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019553.hg.1 |
TC01001273.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019553.hg.1 |
ENST00000324978.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019553.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019553.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019553.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019554.hg.1 |
TC01001273.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019554.hg.1 |
ENST00000324978.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019554.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019554.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010072.hg.1 |
ENST00000612670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010072.hg.1 |
ENST00000612670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173217.hg.1 |
TC0100010074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010072.hg.1 |
ENST00000612670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010072.hg.1 |
ENST00000612670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173218.hg.1 |
TC0100010074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010074.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000324978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000355197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
ENST00000484864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
NM_080429.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173216.hg.1 |
TC0100010074.hg.1 |
Clariom D TC |