Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278648.1 | NP_001265577.1 | 1 - 2 | 76 | 84 | |
XM_017001113.1 | XP_016856602.1 | 1 - 2 | 73 | 84 | ||
XM_017001114.1 | XP_016856603.1 | 1 - 2 | 60 | 84 | ||
GenBank mRNA | AK312786.1 | - | 1 - 2 | 84 | ||
BP281477.1 | - | 1 - 2 | 64 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |