Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000337198.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000347770.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000518940.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000520402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000521536.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000522311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000523071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000523071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
ENST00000523071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_001301668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_015878.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012447.hg.1 |
NM_148174.3 |
Refseq |