Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000581.hg.1 |
TC06000061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000581.hg.1 |
ENST00000342415.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000581.hg.1 |
ENST00000397475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000581.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147053.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147053.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147053.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147053.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147053.hg.1 |
TC0600006870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000342415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
ENST00000496946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006870.hg.1 |
NM_152551.3 |
Refseq |