Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174894.hg.1 |
TC1700009648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174895.hg.1 |
TC1700009648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000250055.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000538513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
ENST00000570788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009648.hg.1 |
NM_006942.1 |
Refseq |