Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001048.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001048.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001048.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001048.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145873.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |