Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003108.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003108.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003108.hg.1 |
TC11000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003109.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003109.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003109.hg.1 |
TC11000235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000529793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000529793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000302797.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
ENST00000526914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010284.hg.1 |
NM_147199.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149188.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149188.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149188.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149188.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149188.hg.1 |
TC1100006988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000529793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000529793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006988.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
ENST00000314254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
ENST00000618161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
NM_054032.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149189.hg.1 |
TC1100006988.hg.1 |
Clariom D TC |