Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000355377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000562711.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000565183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000566261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000568433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
ENST00000570264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_001308157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011360.hg.1 |
NM_016524.3 |
Refseq |