Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011512262.2 | XP_011510564.1 | 4 - 5 | 711 | 74 | |
XM_017005474.1 | XP_016860963.1 | 4 - 5 | 711 | 74 | ||
XM_017005475.1 | XP_016860964.1 | 4 - 5 | 711 | 74 | ||
XM_017005476.1 | XP_016860965.1 | 5 - 6 | 741 | 74 | ||
XM_017005477.1 | XP_016860966.1 | 5 - 6 | 2336 | 74 | ||
XM_017005478.1 | XP_016860967.1 | 5 - 6 | 467 | 74 | ||
GenBank mRNA | BC041437.1 | - | 5 - 6 | 444 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |