Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013386.hg.1 |
TC08001052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013386.hg.1 |
ENST00000221132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013386.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167049.hg.1 |
TC0800009875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000221132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000519862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000524158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
ENST00000613472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009875.hg.1 |
NM_003844.3 |
Refseq |