Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199188.2 | NP_954658.2 | 1 - 2 | 216 | 107 | |
XM_011537831.2 | XP_011536133.1 | 1 - 2 | 197 | 107 | ||
XM_017018755.1 | XP_016874244.1 | 1 - 2 | 130 | 107 | ||
GenBank mRNA | AF370416.1 | - | 1 - 2 | 97 | 107 | |
AK055521.1 | - | 1 - 2 | 61 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |