Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004982.hg.1 |
ENST00000322861.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004982.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004982.hg.1 |
TC16000345.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154351.hg.1 |
TC1600007464.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000322861.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000563728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000566955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
ENST00000568749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007464.hg.1 |
NM_013292.4 |
Refseq |