Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172238.3 | NP_758438.2 | 2 | 864 | 60 | |
GenBank mRNA | AY028376.1 | - | 2 | 864 | 60 | |
DT221015.1 | - | 2 | 566 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |