Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201338.1 | NP_001188267.1 | 17 - 18 | 2505 | 125 | |
NM_001201339.1 | NP_001188268.1 | 17 - 18 | 2502 | 125 | ||
NM_001201340.1 | NP_001188269.1 | 16 - 17 | 2298 | 125 | ||
NM_001320571.1 | NP_001307500.1 | 15 - 16 | 2230 | 125 | ||
NM_001320572.1 | NP_001307501.1 | 17 - 18 | 2502 | 125 | ||
NM_002967.3 | NP_002958.2 | 17 - 18 | 2505 | 125 | ||
XM_017027114.1 | XP_016882603.1 | 15 - 16 | 2187 | 125 | ||
XM_017027115.1 | XP_016882604.1 | 15 - 16 | 2187 | 125 | ||
XM_017027116.1 | XP_016882605.1 | 16 - 17 | 1895 | 125 | ||
XM_017027117.1 | XP_016882606.1 | 16 - 17 | 1892 | 125 | ||
XM_017027118.1 | XP_016882607.1 | 16 - 17 | 1892 | 125 | ||
GenBank mRNA | AK095736.1 | - | 13 - 14 | 1681 | 125 | |
AK290444.1 | - | 17 - 18 | 2398 | 125 | ||
AK293152.1 | - | 14 - 15 | 2003 | 125 | ||
AK294671.1 | - | 15 - 16 | 1955 | 125 | ||
AK295298.1 | - | 14 - 15 | 2088 | 125 | ||
AK298707.1 | - | 16 - 17 | 2194 | 125 | ||
AK299525.1 | - | 17 - 18 | 2263 | 125 | ||
AK304474.1 | - | 13 - 14 | 1653 | 125 | ||
BC126219.1 | - | 17 - 18 | 2376 | 8991935 | 125 | |
BC143937.1 | - | 17 - 18 | 2373 | 9052455 | 125 | |
BC143938.1 | - | 17 - 18 | 2373 | 9052456 | 125 | |
BC143939.1 | - | 17 - 18 | 2373 | 9052457 | 125 | |
L43631.1 | - | 17 - 18 | 2182 | 125 | ||
U72355.1 | - | 17 - 18 | 2387 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |