Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282873.1 | NP_001269802.1 | 44 - 45 | 6741 | 118 | |
NM_015902.5 | NP_056986.2 | 44 - 45 | 6741 | 118 | ||
XM_005250962.4 | XP_005251019.1 | 23 - 24 | 3502 | 118 | ||
XM_011517104.2 | XP_011515406.2 | 43 - 44 | 6726 | 118 | ||
XM_011517105.2 | XP_011515407.2 | 43 - 44 | 6726 | 118 | ||
XM_011517106.2 | XP_011515408.2 | 43 - 44 | 6708 | 118 | ||
XM_017013533.1 | XP_016869022.1 | 43 - 44 | 6726 | 118 | ||
XM_017013534.1 | XP_016869023.1 | 43 - 44 | 6726 | 118 | ||
GenBank mRNA | AB020703.3 | - | 44 - 45 | 6741 | 118 | |
AF006010.2 | - | 44 - 45 | 6319 | 118 | ||
BC137234.1 | - | 44 - 45 | 6313 | 9021231 | 118 | |
U95000.1 | - | 44 - 45 | 6283 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |