Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005515.hg.1 |
TC06000515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005515.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005515.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005516.hg.1 |
TC06000515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005516.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005516.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154054.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154054.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154054.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154054.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154054.hg.1 |
TC0600007799.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154055.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154055.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154055.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154055.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154055.hg.1 |
TC0600007799.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007799.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154056.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154056.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154056.hg.1 |
ENST00000229769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154056.hg.1 |
NM_021922.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154056.hg.1 |
TC0600007799.hg.1 |
Clariom D TC |