Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001442.hg.1 |
ENST00000338221.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001442.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001442.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001442.hg.1 |
TC10000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001444.hg.1 |
ENST00000338221.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001444.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001444.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001444.hg.1 |
TC10000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147277.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147277.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147277.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147277.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147277.hg.1 |
TC1000006911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000147278.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147278.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147278.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147278.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147278.hg.1 |
TC1000006911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000147279.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147279.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147279.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147279.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147279.hg.1 |
TC1000006911.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
ENST00000377495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006911.hg.1 |
NM_001098844.2 |
Refseq |