Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184774.1 | NP_001171703.1 | 13 - 14 | 2995 | 76 | |
NM_001184776.1 | NP_001171705.1 | 12 - 13 | 2803 | 76 | ||
GenBank mRNA | AB023144.2 | - | 12 - 13 | 2763 | 76 | |
BC143851.1 | - | 13 - 14 | 2894 | 9052367 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |