Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024118.hg.1 |
ENST00000343958.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024118.hg.1 |
ENST00000546711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024118.hg.1 |
NM_018351.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024118.hg.1 |
TC12001829.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000343958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000451107.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000546711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000549499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
NM_018351.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000343958.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000451107.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000546711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
ENST00000549499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
NM_018351.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189723.hg.1 |
TC1200011574.hg.1 |
Clariom D TC |