Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011163.hg.1 |
ENST00000262866.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011163.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011163.hg.1 |
TC20000815.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000163079.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163079.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000163079.hg.1 |
TC2000009013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000163079.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000163079.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000262866.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
ENST00000360672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_032214.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009013.hg.1 |
NM_175077.2 |
Refseq |