Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025564.hg.1 |
TC07001662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025564.hg.1 |
ENST00000292377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025564.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025563.hg.1 |
TC07001662.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025563.hg.1 |
ENST00000292377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025563.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185947.hg.1 |
TC0700011979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000480087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000486702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011979.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000292377.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000471717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
ENST00000490629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
NM_152742.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700185946.hg.1 |
TC0700011979.hg.1 |
Clariom D TC |