Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007537.hg.1 |
ENST00000371413.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007537.hg.1 |
ENST00000371418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007537.hg.1 |
NM_005097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007537.hg.1 |
TC10000668.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000371413.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000371418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000485458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000629035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000630184.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000630487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
NM_001308275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
NM_005097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000371413.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000371418.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000485458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000629035.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000630184.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
ENST00000630487.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
NM_001308275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
NM_005097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157749.hg.1 |
TC1000008489.hg.1 |
Clariom D TC |