Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023408.hg.1 |
TC04001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023408.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023408.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023406.hg.1 |
TC04001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023406.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023406.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023406.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023405.hg.1 |
TC04001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023405.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023405.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023407.hg.1 |
TC04001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023407.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023407.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023407.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023409.hg.1 |
TC04001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023409.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023409.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184131.hg.1 |
TC0400012257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184133.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184133.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184133.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184133.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184133.hg.1 |
TC0400012257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000505647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012257.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
ENST00000508011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184132.hg.1 |
TC0400012257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184130.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184130.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184130.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184130.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184130.hg.1 |
TC0400012257.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184135.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184135.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184135.hg.1 |
ENST00000434826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184135.hg.1 |
NM_152543.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184135.hg.1 |
TC0400012257.hg.1 |
Clariom D TC |