Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017130.hg.1 |
TC04001259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017130.hg.1 |
ENST00000344157.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017130.hg.1 |
ENST00000355665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017130.hg.1 |
NM_001031732.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017130.hg.1 |
NM_133370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR4_ctg9_hap1000035.hg.1 |
TC4_ctg9_hap1000003.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR4_ctg9_hap1000035.hg.1 |
NM_001031732.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR4_ctg9_hap1000035.hg.1 |
NM_133370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000344157.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000355665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000579690.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
NM_001031732.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
NM_133370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000344157.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000355665.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
ENST00000579690.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
NM_001031732.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
NM_133370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174187.hg.1 |
TC0400010904.hg.1 |
Clariom D TC |