Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029214.hg.1 |
TC06002330.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029214.hg.1 |
ENST00000366787.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029214.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000435791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000472733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000488355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013998.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000366787.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000461848.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
ENST00000617924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
NM_003247.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196418.hg.1 |
TC0600013998.hg.1 |
Clariom D TC |