Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012421.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012421.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012421.hg.1 |
TC11000998.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012420.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012420.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012420.hg.1 |
TC11000998.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166761.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166761.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166761.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100166761.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100166761.hg.1 |
TC1100009026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000304987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
ENST00000533868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009026.hg.1 |
NM_015191.2 |
Refseq |