Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000654.hg.1 |
TC0X000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000654.hg.1 |
ENST00000304087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000654.hg.1 |
ENST00000380682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000654.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000655.hg.1 |
TC0X000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000655.hg.1 |
ENST00000304087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000655.hg.1 |
ENST00000380682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000655.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145275.hg.1 |
TC0X00006616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006616.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
ENST00000380682.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
ENST00000616992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
NM_014728.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145274.hg.1 |
TC0X00006616.hg.1 |
Clariom D TC |