Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014652.hg.1 |
TC01000933.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014652.hg.1 |
ENST00000271311.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014652.hg.1 |
ENST00000370017.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014652.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014651.hg.1 |
TC01000933.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014651.hg.1 |
ENST00000271311.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014651.hg.1 |
ENST00000370017.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014651.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166445.hg.1 |
TC0100009329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100166444.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166444.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166444.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166444.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166444.hg.1 |
TC0100009329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000370017.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
ENST00000445274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009329.hg.1 |
NM_001144937.1 |
Refseq |