Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
ENST00000249837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
ENST00000261517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
ENST00000395896.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
ENST00000395898.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
NM_001018088.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
NM_017684.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
NM_018080.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
NM_020821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014355.hg.1 |
TC15001510.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000249837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000261517.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000395896.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000395898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_001018088.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_017684.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_018080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_020821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000249837.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000261517.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000395896.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
ENST00000395898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_001018088.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_017684.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_018080.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
NM_020821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171329.hg.1 |
TC1500009669.hg.1 |
Clariom D TC |