Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291661.1 | NP_001278590.1 | 62 - 63 | 10168 | 143 | |
XM_011533466.1 | XP_011531768.1 | 62 - 63 | 10153 | 143 | ||
XM_011533467.1 | XP_011531769.1 | 62 - 63 | 10120 | 143 | ||
XM_011533468.1 | XP_011531770.1 | 62 - 63 | 10090 | 143 | ||
XM_011533469.1 | XP_011531771.1 | 61 - 62 | 10012 | 143 | ||
XM_017005860.1 | XP_016861349.1 | 62 - 63 | 10153 | 143 | ||
XM_017005861.1 | XP_016861350.1 | 61 - 62 | 10836 | 143 | ||
XM_017005862.1 | XP_016861351.1 | 41 - 42 | 6728 | 143 | ||
GenBank mRNA | AK097746.1 | - | 3 - 4 | 454 | 143 | |
AK128592.1 | - | 10 - 11 | 1594 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |