Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291661.1 | NP_001278590.1 | 66 - 67 | 10852 | 140 | |
XM_011533466.1 | XP_011531768.1 | 66 - 67 | 10837 | 140 | ||
XM_011533467.1 | XP_011531769.1 | 66 - 67 | 10804 | 140 | ||
XM_011533468.1 | XP_011531770.1 | 66 - 67 | 10774 | 140 | ||
XM_011533469.1 | XP_011531771.1 | 65 - 66 | 10696 | 140 | ||
XM_017005860.1 | XP_016861349.1 | 66 - 67 | 10837 | 140 | ||
XM_017005861.1 | XP_016861350.1 | 65 - 66 | 11520 | 140 | ||
XM_017005862.1 | XP_016861351.1 | 45 - 46 | 7412 | 140 | ||
GenBank mRNA | AK128592.1 | - | 14 - 15 | 2278 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |