Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000525.hg.1 |
ENST00000270586.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000525.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000525.hg.1 |
TC17000052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000526.hg.1 |
ENST00000270586.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000526.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000526.hg.1 |
TC17000052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000575643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006644.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145470.hg.1 |
TC1700006644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000270586.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000571309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000575079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
ENST00000614486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
NM_001270481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
NM_002798.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145471.hg.1 |
TC1700006644.hg.1 |
Clariom D TC |