Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035362.hg.1 |
TC01002308.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035362.hg.1 |
ENST00000375105.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035362.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013167.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197414.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197414.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197414.hg.1 |
TC0100013167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197419.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197419.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197419.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197419.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197419.hg.1 |
TC0100013167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197416.hg.1 |
TC0100013167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
ENST00000375105.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
ENST00000617227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
NM_001271814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
NM_012400.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197417.hg.1 |
TC0100013167.hg.1 |
Clariom D TC |