Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006420.2 | NP_006411.2 | 34 - 35 | 4775 | 63 | |
XM_005260252.3 | XP_005260309.1 | 34 - 35 | 4789 | 63 | ||
GenBank mRNA | AB209397.1 | - | 14 - 15 | 1731 | 63 | |
AF084521.1 | - | 34 - 35 | 4623 | 63 | ||
AK022623.1 | - | 13 - 14 | 1597 | 63 | ||
AK074303.1 | - | 17 - 18 | 2151 | 63 | ||
AK302526.1 | - | 20 - 21 | 2630 | 63 | ||
BC144625.1 | - | 33 - 34 | 4805 | 63 | ||
BC144626.1 | - | 35 - 36 | 5560 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |