Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000926.4 | NP_000917.3 | 6 - 7 | 3235 | 153 | |
NM_001202474.3 | NP_001189403.1 | 6 - 7 | 2485 | 153 | ||
NM_001271161.2 | NP_001258090.1 | 5 - 6 | 2179 | 153 | ||
NM_001271162.1 | NP_001258091.1 | 6 - 7 | 1155 | 153 | ||
NR_073141.2 | 6 - 7 | 2433 | 153 | |||
NR_073142.2 | 5 - 6 | 2316 | 153 | |||
GenBank mRNA | AB084248.1 | - | 5 - 6 | 2186 | 153 | |
AB085843.1 | - | 5 - 6 | 2323 | 153 | ||
AB085845.1 | - | 6 - 7 | 2440 | 153 | ||
AF016381.1 | - | 6 - 7 | 2667 | 153 | ||
AK304853.1 | - | 6 - 7 | 1155 | 153 | ||
AY212933.1 | - | 3 - 4 | 1865 | 153 | ||
M15716.1 | - | 6 - 7 | 2667 | 153 | ||
X51730.1 | - | 6 - 7 | 3946 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons |