Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055960.hg.1 |
TC01003638.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055960.hg.1 |
ENST00000367468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055960.hg.1 |
ENST00000490885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055960.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226820.hg.1 |
TC0100016715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000367468.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000466691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000490885.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
ENST00000559800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016715.hg.1 |
NM_000963.3 |
Refseq |