Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
ENST00000312777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
ENST00000405876.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011052.hg.1 |
TC12000853.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201540.hg.1 |
TC1200012704.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000312777.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000405876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000536408.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000537880.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000549550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
ENST00000551627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_001143852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012704.hg.1 |
NM_032300.4 |
Refseq |