Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019024.2 | NP_061897.1 | 26 - 27 | 4198 | 82 | |
XM_006712034.2 | XP_006712097.1 | 26 - 27 | 4137 | 82 | ||
XM_006712035.3 | XP_006712098.1 | 26 - 27 | 4201 | 82 | ||
XM_011532934.2 | XP_011531236.1 | 26 - 27 | 4665 | 82 | ||
XM_011532935.2 | XP_011531237.1 | 15 - 16 | 2337 | 82 | ||
XM_017004378.1 | XP_016859867.1 | 26 - 27 | 4137 | 82 | ||
XM_017004379.1 | XP_016859868.1 | 27 - 28 | 4575 | 82 | ||
XM_017004380.1 | XP_016859869.1 | 27 - 28 | 4348 | 82 | ||
GenBank mRNA | AB037835.1 | - | 17 - 18 | 2645 | 82 | |
AK026928.1 | - | 3 - 4 | 458 | 82 | ||
AK096637.1 | - | 26 - 27 | 4137 | 82 | ||
BC150605.1 | - | 26 - 27 | 4183 | 9056975 | 82 | |
BX538008.1 | - | 20 - 21 | 3121 | 82 | ||
BX648805.1 | - | 12 - 13 | 2358 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |