Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012391.hg.1 |
TC06004099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012391.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012391.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199602.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199602.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199602.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199602.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199602.hg.1 |
TC0600014189.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
ENST00000367576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014189.hg.1 |
NM_032860.4 |
Refseq |