Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
TC07000454.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
ENST00000223398.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
ENST00000361545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
ENST00000395060.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
ENST00000537700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
ENST00000539676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
NM_003388.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006171.hg.1 |
NM_032421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000223398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000361545.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000395060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
NM_003388.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
NM_032421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000223398.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000361545.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
ENST00000395060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
NM_003388.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
NM_032421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155316.hg.1 |
TC0700008003.hg.1 |
Clariom D TC |