Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000297494.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000460603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000460603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000460603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000461406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000467517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000468293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000473057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000473057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000473057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000473057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000475454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000477227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
ENST00000484524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_000603.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009688.hg.1 |
NM_001160111.1 |
Refseq |