Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000094.3 | NP_000085.1 | 70 - 71 | 5822 | 160 | |
XM_011533337.1 | XP_011531639.1 | 71 - 72 | 5857 | 160 | ||
XM_017005688.1 | XP_016861177.1 | 71 - 72 | 5857 | 160 | ||
XM_017005689.1 | XP_016861178.1 | 71 - 72 | 5857 | 160 | ||
XM_017005690.1 | XP_016861179.1 | 71 - 72 | 5857 | 160 | ||
XM_017005691.1 | XP_016861180.1 | 71 - 72 | 5857 | 160 | ||
XM_017005692.1 | XP_016861181.1 | 71 - 72 | 5857 | 160 | ||
GenBank mRNA | AB209645.1 | - | 36 - 37 | 1738 | 160 | |
L02870.1 | - | 70 - 71 | 5934 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |