Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031698.2 | NP_001026868.2 | 23 - 24 | 2555 | 168 | |
NM_012272.2 | NP_036404.1 | 22 - 23 | 2827 | 168 | ||
XM_006719324.3 | XP_006719387.1 | 22 - 23 | 3612 | 168 | ||
XM_006719325.3 | XP_006719388.1 | 22 - 23 | 3609 | 168 | ||
XM_011538135.2 | XP_011536437.1 | 22 - 23 | 3612 | 168 | ||
XM_011538136.2 | XP_011536438.1 | 22 - 23 | 3609 | 168 | ||
XM_011538137.2 | XP_011536439.1 | 21 - 22 | 3531 | 168 | ||
XM_011538138.2 | XP_011536440.1 | 21 - 22 | 3501 | 168 | ||
XM_011538139.2 | XP_011536441.1 | 23 - 24 | 2552 | 168 | ||
XM_011538140.2 | XP_011536442.1 | 23 - 24 | 2549 | 168 | ||
XM_011538141.1 | XP_011536443.1 | 22 - 23 | 2378 | 168 | ||
XM_011538143.2 | XP_011536445.1 | 22 - 23 | 2394 | 168 | ||
XM_011538144.2 | XP_011536446.1 | 21 - 22 | 2267 | 168 | ||
XM_017019135.1 | XP_016874624.1 | 22 - 23 | 2315 | 168 | ||
XM_017019136.1 | XP_016874625.1 | 21 - 22 | 2264 | 168 | ||
XM_017019137.1 | XP_016874626.1 | 21 - 22 | 2264 | 168 | ||
XM_017019139.1 | XP_016874628.1 | 18 - 19 | 2278 | 168 | ||
GenBank mRNA | AK027117.1 | - | 8 - 9 | 836 | 168 | |
AL137459.1 | - | 9 - 10 | 1058 | 168 | ||
AL834216.1 | - | 22 - 23 | 2827 | 168 | ||
BC031917.1 | - | 20 - 21 | 2151 | 168 | ||
BC067364.1 | - | 22 - 23 | 2379 | 6644672 | 168 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |