Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007259.hg.1 |
TC0X000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007259.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007259.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007259.hg.1 |
ENST00000540483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007259.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007257.hg.1 |
TC0X000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007257.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007257.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007261.hg.1 |
TC0X000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007261.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007261.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007261.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007260.hg.1 |
TC0X000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007260.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007260.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007260.hg.1 |
ENST00000540483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007260.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155522.hg.1 |
TC0X00008120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008120.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155518.hg.1 |
TC0X00008120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155520.hg.1 |
TC0X00008120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000465301.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000520821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
ENST00000540313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
NM_020769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155521.hg.1 |
TC0X00008120.hg.1 |
Clariom D TC |