Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006317.hg.1 |
TC09000568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006317.hg.1 |
ENST00000374232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006317.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156577.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156577.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156577.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156577.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156577.hg.1 |
TC0900008474.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000416585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
ENST00000604751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008474.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156578.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156578.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156578.hg.1 |
ENST00000374232.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156578.hg.1 |
NM_001012994.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156578.hg.1 |
TC0900008474.hg.1 |
Clariom D TC |