Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021073.hg.1 |
ENST00000293406.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021073.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021073.hg.1 |
TC17001571.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010810.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000293406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000585388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000590563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
ENST00000591247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
NM_152344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186330.hg.1 |
TC1700010810.hg.1 |
Clariom D TC |