Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017666.4 | NP_060136.1 | 12 - 13 | 1597 | 85 | |
GenBank mRNA | AK000102.1 | - | 12 - 13 | 1520 | 85 | |
AK290373.1 | - | 12 - 13 | 1548 | 85 | ||
AL834333.1 | - | 12 - 13 | 1528 | 85 | ||
BC051728.1 | - | 12 - 13 | 1548 | 4828435 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |