Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270546.1 | NP_001257475.1 | 26 - 27 | 3620 | 114 | |
NM_006738.5 | NP_006729.4 | 34 - 35 | 7821 | 114 | ||
NM_007200.4 | NP_009131.2 | 34 - 35 | 7809 | 114 | ||
GenBank mRNA | AB055890.1 | - | 35 - 36 | 7828 | 114 | |
AB209414.1 | - | 23 - 24 | 3102 | 114 | ||
AF126008.1 | - | 26 - 27 | 3463 | 114 | ||
AF387101.1 | - | 33 - 34 | 7615 | 114 | ||
AF406992.1 | - | 34 - 35 | 7708 | 114 | ||
BC063592.1 | - | 21 - 22 | 2596 | 114 | ||
BC140862.1 | - | 19 - 20 | 2432 | 114 | ||
BC171798.1 | - | 19 - 20 | 2432 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |