Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
TC03001303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022465.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022463.hg.1 |
TC03001303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022463.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022463.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022463.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022463.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022464.hg.1 |
TC03001303.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022464.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022464.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022464.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022464.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010772.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178543.hg.1 |
TC0300010772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178544.hg.1 |
TC0300010772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000340369.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000396251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
ENST00000421646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
NM_001198621.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
NM_194293.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178545.hg.1 |
TC0300010772.hg.1 |
Clariom D TC |