Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142349.1 | NP_001135821.1 | 2 - 3 | 642 | 168 | |
NM_174934.3 | NP_777594.1 | 3 - 4 | 705 | 168 | ||
GenBank mRNA | AK314531.1 | - | 3 - 4 | 618 | 168 | |
AY149967.1 | - | 3 - 4 | 616 | 168 | ||
BC035017.2 | - | 2 - 3 | 606 | 4817294 | 168 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |